F5398
FK-Binding Protein human
recombinant, expressed in E. coli
Synonym(s):
FKBP-12
About This Item
recombinant
expressed in E. coli
UniProt accession no.
storage temp.
2-8°C
Gene Information
human ... FKBP1A(2280)
General description
Biochem/physiol Actions
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service