Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F4129

Sigma-Aldrich

Fibrinogen from human plasma

55-75% protein, ≥60% of protein is clottable

Synonym(s):

Factor I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

biological source

human plasma

form

powder

quality

≥60% of protein is clottable

mol wt

α-chain 63.5 kDa
β-chain 56 kDa
γ chain 47 kDa (about 4% carbohydrate content)
soluble dimer 340 kDa

concentration

55-75% protein (biuret)
55-75% protein

solubility

0.9% NaCl: soluble 10 mg/mL

UniProt accession no.

Storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Fibrinogen, or Factor I, is a blood protein that is involved in clotting and is converted to fibrin by thrombin. Fibrinogen has an approximate molecular weight of 340 kDa.

Application

Fibrinogen was used in velocity variation assessment of red blood cell aggregation using spectral domain Doppler optical coherence tomography.[1] It was also used to study the nano-bio interaction between graphite oxide nanoparticles and human blood components.[2]

Biochem/physiol Actions

Acute phase protein that is part of the coagulation cascade of proteins.
Fibrinogen is an acute phase protein that is part of the coagulation cascade of proteins. The end result of the cascade is the production of thrombin that converts fibrinogen to fibrin. Thrombin rapidly proteolyses fibrinogen, releasing fibrinopeptide A. The loss of this small peptide is not sufficient to make the resulting fibrin molecule insoluble, but it tends to form complexes with adjacent fibrin and fibrinogen molecules. Thrombin then cleaves a second peptide, fibrinopeptide B, from fibrin and the fibrin monomers formed then polymerize spontaneously to form an insoluble gel. The polymerized fibrin is held together by noncovalent and electrostatic forces and stabilized by the transamidating enzyme, factor XIIIa, that is produced by the action of thrombin on factor XIII. The insoluble fibrin aggregates (clots) and aggregated platelets then block the damaged blood vessel and prevent further bleeding. The amount of fibrinogen in the plasma can serve as a nonspecific indicator of whether or not an inflammatory process is present in the body. Fibrinogen from any mammalian source will be cleaved by thrombin from any mammalian source.

Specifications

Source material has tested negative for HIV and HBsAg.

Physical form

Lyophilized powder containing ~15% sodium citrate and ~20% sodium chloride.

Analysis Note

Protein determined by biuret

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nano-Bio Interaction between Graphite Oxide Nano Particles and Human Blood Components
Hyoung-Mi Kim, Kyoung-Min Kim, Kyunghong Lee, Yoon Suk Kim and Jae-Min Oh
European Journal of Organic Chemistry, 5343-5349 (2012)
[The number of bacteria and fungi in "da-shan-zha-wan" pills].
Y Zhou
Zhong yao tong bao (Beijing, China : 1981), 10(12), 27-28 (1985-12-01)
Sachie Hiratsuka et al.
EMBO molecular medicine, 10(7) (2018-06-23)
Primary tumours establish metastases by interfering with distinct organs. In pre-metastatic organs, a tumour-friendly microenvironment supports metastatic cells and is prepared by many factors including tissue resident cells, bone marrow-derived cells and abundant fibrinogen depositions. However, other components are unclear.
Katarina Waldén et al.
The Annals of thoracic surgery, 97(4), 1199-1206 (2014-02-11)
Data from small selected patient populations suggest that the preoperative plasma concentration of fibrinogen influences postoperative blood loss and red blood cell transfusion after cardiac operations, but there are also conflicting reports. We assessed the importance of preoperative fibrinogen concentration
Richard J Fish et al.
Blood, 123(14), 2278-2281 (2014-02-21)
Mutations in the human fibrinogen genes can lead to the absence of circulating fibrinogen and cause congenital afibrinogenemia. This rare bleeding disorder is associated with a variable phenotype, which may be influenced by environment and genotype. Here, we present a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service