Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

F406-05

Sigma-Aldrich

Feline Osteoblasts: FOb (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

feline bone (normal)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 38

morphology

osteoblast

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

arthritis; osteoporosis

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.Feline Osteoblasts (FOb), provide a model system to study matrix mineralization, effects of inflammatory mediators and bone morphogenic proteins, physiological control of bone remodeling and regulation of bone metabolism.Characterization: Positive for bone mineralization.

Cell Line Origin

Bone

Application

model for skeletal system research, signaling pathways, bone morphogenesis, gene expression, cell activation and proliferation, wound healing, bone formation and regeneration, treatment strategies, tissue engineering, in vitro bone models, mineralization, hormone treatments

Components

Feline Osteoblast Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Feline Osteoblast Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the FOb Culture Protocol.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service