Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F3803

Sigma-Aldrich

Monoclonal Anti-Follicular Dendritic Cells (FDC) antibody produced in mouse

clone CNA.42, tissue culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

CNA.42, monoclonal

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:50 using human tonsil
immunohistochemistry (frozen sections): 1:25-1:50 using acetone-fixed, cryostat sections of human tonsil

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Immunogen

mouse CEM T cell line.

Application

Monoclonal Anti-Follicular Dendritic Cells (FDC) antibody is suitable for immunohistochemistry studies of formalin-fixed, paraffin-embedded sections at a dilution of 1:25-1:50 using human tonsil and frozen sections at a dilution of 1:25-1:50 using acetone-fixed, cryostat sections of human tonsil.

Biochem/physiol Actions

Follicular dendritic cells (FDCs) are phenotypically distinct cells that are restricted to primary and secondary follicles of any secondary lymphoid organs, lymphoid dendritic cells are found in T cell-rich interfollicular areas. FDCs are morphologically large cells (often containing multiple nuclei) that exhibit numerous dendritic processes. Initially, FDCs are characterized as ′accessory′ cells that passively support germinal centre (GC) responses. However, latter it has been postulated that FDCs actively shape humoral immunity.

Physical form

Tissue culture supernatant (culture medium with fetal calf serum), containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balthasar A Heesters et al.
Nature reviews. Immunology, 14(7), 495-504 (2014-06-21)
Follicular dendritic cells (FDCs) are essential for high-affinity antibody production and for the development of B cell memory. Historically, FDCs have been characterized as 'accessory' cells that passively support germinal centre (GC) responses. However, recent observations suggest that FDCs actively
S Petrasch et al.
British journal of haematology, 80(1), 21-26 (1992-01-01)
Follicular dendritic cells (FDC) are located within the B-cell follicles of non-malignant lymphatic tissues and within non-Hodgkin-lymphomas (NHL) derived from the germinal centre or the mantlezone. The interactions between FDC and non-neoplastic B-cells have been extensively investigated but so far
Kirsten van Nierop et al.
Seminars in immunology, 14(4), 251-257 (2002-08-07)
Follicular dendritic cells (FDCs) have important functions in the selection of memory B lymphocytes during germinal center reactions (GCR). They present native antigens to potential memory cells, of which only B cells with high affinity B cell receptors (BCR) can
Lili Wang et al.
Human gene therapy, 22(11), 1389-1401 (2011-04-12)
Vectors based on the primate-derived adeno-associated virus serotype 8 (AAV8) are being evaluated in preclinical and clinical models. Natural infections with related AAVs activate memory B cells that produce antibodies capable of modulating the efficacy and safety of the vector.
Eirini Moysi et al.
Frontiers in immunology, 12, 683396-683396 (2021-06-29)
CD4 T cells are key mediators of adaptive immune responses during infection and vaccination. Within secondary lymphoid organs, helper CD4 T cells, particularly those residing in germinal centers known as follicular helper T cells (Tfh), provide critical help to B-cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service