Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F3179

Sigma-Aldrich

Anti-Frizzled-3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FZD3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5 μg/mL using human skeletal muscle

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD3(7976)
mouse ... Fzd3(14365)
rat ... Fzd3(266715)

General description

FZD3 belongs to the frizzled (Fz) family. Frizzled homolog protein (FZD) is a seven-pass transmembrane type receptor expressed in various normal tissues (skeletal muscle, kidney, pancreas, cerebellum and cerebral cortex). In human, ten members (FZD1-FZD10) of Frizzled homolog protein have been identified. It is expressed in several cancers (esophageal carcinoma, lung squamous cell carcinoma, primary acute and lymphoblastic leukemia, myeloma, lymphoma and Ewing sarcoma).[1]

Immunogen

synthetic peptide corresponding to the N-terminal extracellular domain of human frizzled-3, conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Application

Anti-Frizzled-3 antibody is suitable for western blot analysis. It is also suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 5μg/mL, using human skeletal muscle.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S B Wang et al.
Genetics and molecular research : GMR, 15(1) (2016-02-26)
Frizzled 3 is an important receptor in the Wnt/β-catenin pathway, a conserved signaling pathway that regulates gene expression and controls diverse developmental processes. However, the role of this protein during follicular development in the adult ovary is not known. The
Yoshimi Endo et al.
Molecular and cellular biology, 28(7), 2368-2379 (2008-01-24)
Recombinant Wnt-3a stimulated the rapid formation of elongated processes in Ewing sarcoma family tumor (ESFT) cells that were identified as neurites. The processes stained positively for polymerized actin and microtubules as well as synapsin I and growth-associated protein 43. Inhibition
T Nohno et al.
Cellular and molecular biology (Noisy-le-Grand, France), 45(5), 653-659 (1999-10-08)
Members of the frizzled (Fz) family are involved in Wnt signaling during embryogenesis in the vertebrate. We identified chicken cognates of Fz-2, Fz-3, Fz-4, Fz-6 and Fz-8, and examined spatial and temporal expression patterns in the chick embryos by whole-mount
Sze Chuen Cesar Wong et al.
PloS one, 8(11), e79481-e79481 (2013-11-21)
Frizzled homolog 3 receptor was up-regulated in several gastrointestinal cancers such as esophageal and gastric cancers. Moreover, frizzled homolog 3 has recently reported to be expressed in colorectal adenoma specimens. In the present study, we investigated the clinical significance of

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service