Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1805

Sigma-Aldrich

Anti-Flotillin 2 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ECS1, Anti-ESA, Anti-Epidermal surface antigen, Anti-FLOT2, Anti-Reggie-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using human HEK-293T cells
western blot: 0.5-1 μg/mL using whole extract of mouse 3T3 cells
western blot: 2-4 μg/mL using whole extract of rat PC12 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FLOT2(2319)
mouse ... Flot2(14252)
rat ... Flot2(83764)

General description

Flotillins are lipid raft-associated proteins, and belong to the integral membrane proteins. Flotillins contain an evolutionarily conserved domain called the prohibitin homology (PHB) domain or SPFH domain (stomatin/prohibitin/flotillin/HflK/C domain). The lipid recognition motif present in the PHB domain, is responsible for targeting proteins to the plasma membrane. Flotillin 2 (FLOT2) is widely distributed in human tissues. FLOT2 is mainly present at the plasma membrane, but also localizes in intracellular compartments including lipid droplets and compartments of the endocytic pathway.[1]

Immunogen

synthetic peptide corresponding to amino acids 232-245 of human flotillin 2 with an N-terminal added cysteine, conjugated to KLH. The corresponding sequence is identical in rat and mouse.

Application

Anti-Flotillin 2 antibody is suitable for:
  • indirect immunofluorescence (10-20μg/mL using human HEK-293T cells).
  • western blot (0.5-1μg/mL using whole extract of mouse 3T3 cells).
  • western blot (2-4μg/mL using whole extract of rat PC12 cells).
The antibody has been used for western blotting of proteins isolated from HeLa cells, primary mouse oesophageal epithelial cells and HEK-293 cells.[2]
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Flotillins are scaffold proteins which are crucial for the formation and dynamics of lipid rafts. They are associated in different biological processes, such as endocytosis, phagocytosis, neuronal regeneration, insulin signaling etc. FLOT2 (Flotillin 2) is a target of miR-34a.[3] It is up-regulated in nasopharyngeal carcinoma, renal cell carcinoma, breast cancer and gastric cancer. Knockdown of FLOT2 reduces cell proliferation, migration, and invasion.[4] FLOT2 interacts with γ-catenin and regulates cell-cell adhesion in non-malignant epithelial cells.[5]

Target description

Flotillin 2 encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling. Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuyuan Wen et al.
PloS one, 9(7), e101676-e101676 (2014-07-12)
Nasopharyngeal carcinoma (NPC) is a head and neck malignant tumor rare throughout most of the world but common in Southeast Asia, especially in Southern China. Flotillin-2 (Flot-2) is not only an important component of cellular membrane, but also involves in
Yusuke Amanuma et al.
Scientific reports, 5, 14142-14142 (2015-09-17)
Acetaldehyde is an ethanol-derived definite carcinogen that causes oesophageal squamous cell carcinoma (ESCC). Aldehyde dehydrogenase 2 (ALDH2) is a key enzyme that eliminates acetaldehyde, and impairment of ALDH2 increases the risk of ESCC. ALDH2 is produced in various tissues including
Janelle R Walton et al.
Histochemistry and cell biology, 139(3), 487-500 (2012-10-16)
A proteomics survey of human placental syncytiotrophoblast (ST) apical plasma membranes revealed peptides corresponding to flotillin-1 (FLOT1) and flotillin-2 (FLOT2). The flotillins belong to a class of lipid microdomain-associated integral membrane proteins that have been implicated in clathrin- and caveolar-independent
Anusha et al.
Cell death & disease, 12(3), 259-259 (2021-03-13)
Chronic myeloid leukemia (CML) patients with complex chromosomal translocations as well as non-compliant CML patients often demonstrate short-lived responses and poor outcomes on the current therapeutic regimes using Imatinib and its variants. It has been derived so far that leukemic
A Zachary Ostrow et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(12), E2411-E2419 (2017-03-08)
Forkhead Box (Fox) proteins share the Forkhead domain, a winged-helix DNA binding module, which is conserved among eukaryotes from yeast to humans. These sequence-specific DNA binding proteins have been primarily characterized as transcription factors regulating diverse cellular processes from cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service