Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1777

Sigma-Aldrich

Monoclonal Anti-HLA-DQ−FITC antibody produced in mouse

clone HK19, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-HLA-DQ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HK19, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 10 μL using 1 × 106 cells

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

Storage temp.

2-8°C

Gene Information

human ... HLA-DQB1(3119)

Specificity

Recognizes MHC Class II molecules that are preferentially expressed on the surface of B lymphocytes. The antibody reacts weakly with resting or ConA-activated T cells, monocytes, granulocytes, and erythrocytes.

Immunogen

GM-1500 human tumor cell line.

Target description

HLA-DQ molecules are bimolecular glycoprotein complexes of 29 kDa and 33 kDa components. The HLA-DQ antigens are present on peripheral blood lymphocytes and tonsillar and splenic mononuclear cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Preparation Note

Prepared by conjugation to fluorescein isothiocyanate isomer I (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mahmoudi-Rad et al.
Clinical and experimental dermatology, 38(6), 646-651 (2013-07-11)
Various materials have been investigated as possible skin substitutes to repair skin defects such as burns. Because of its unique characteristics, acellular amniotic membrane seems to provide a good scaffold for cell cultures. To investigate the proliferation of fibroblasts on
H Moravvej et al.
The British journal of dermatology, 179(1), 72-79 (2018-01-14)
Different methods of fibroblast application have been examined to treat recessive dystrophic epidermolysis bullosa (RDEB). To compare the effects of intradermal injection of cultured allogeneic fibroblasts in healing RDEB wounds with those of fibroblasts seeded on amniotic membrane scaffolds (FAMS)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service