Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

F150-05

Sigma-Aldrich

Feline Skeletal Muscle Cells: FSkMC (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

feline skeletal muscle (normal limb)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 38

morphology

Skeletal muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

muscular dystrophy

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Feline Skeletal Muscle Cells (FSkMC) provide a useful system to study many aspects of muscular function and disease and can serve as an intermediate model of human diseases. Skeletal Muscle Cells also play an instrumental role in the glucose metabolism and diabetes.

Cell Line Origin

Muscle

Application

lipid and glucose metabolism, exercise, insulin action, cell signaling

Components

Feline Skeletal Muscle Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Feline Skeletal Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the FSkMC Culture Protocol.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service