Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F139

Sigma-Aldrich

Anti-Fra2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot (chemiluminescent): 10 μg/mL using Fra2 expressed in human HeLa cells

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FOSL2(2355)

Specificity

The antibody reacts specifically with Fra2 (Fos-related antigen 2), a cellular oncogene product expressed in HeLa cells.

Immunogen

synthetic peptide derived from amino acids 288-302 from the C-terminal region of human Fra2.

Physical form

Lyophilized from phosphate buffered saline, containing 0.05% sodium azide.

Reconstitution

Reconstitute with 100 μl deionized water.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service