F1043
Monoclonal Anti-Filensin antibody produced in mouse
clone FIL-7B10, ascites fluid
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
FIL-7B10, monoclonal
mol wt
antigen 115 kDa
contains
15 mM sodium azide
species reactivity
bovine, sheep, human
should not react with
mouse, chicken, rat
technique(s)
immunohistochemistry (frozen sections): suitable
indirect immunofluorescence: 1:400 using bovine lens frozen sections
microarray: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... BFSP1(631)
Specificity
Immunogen
Application
Western Blotting (1 paper)
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service