Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F0675

Sigma-Aldrich

Anti-Fractalkine antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CX3CL1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5-15 μg/mL using human tissue
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CX3CL1(6376)

General description

Fractalkine, also known as neurotactin is an atypical chemokine, synthesized as a membrane-bound molecule containing a chemokine domain presented on a mucin-like stalk which facilitates the direct capture of circulating leukocytes. It also has a crucial role in all stages of atherogenesis like recruitment, retention and survival of monocytes in early atherosclerotic lesions along with the foam cell persistence in advanced plaques. Anti-Fractalkine antibody can be used in neutralization of biological activity of recombinant human fractalkine. It can also be used in immunohistochemistry. Goat anti-Fractalkine antibody has shown 35% (approx.) cross-reactivity with recombinant mouse fractalkine/CX3CL1 and 15% (approx.) cross-reactivity with recombinant rat CX3CL1. This antibody also has 5% cross-reacitivity with recombinant human eotaxin, recombinant human MCP-1, recombinant mouse MARC, recombinant human MCP-2, recombinant human 6Ckine and recombinant mouse eotaxin.

Specificity

The antibody will neutralize the biological activity of recombinant human fractalkine/CX3CL1. Based on ELISA, the antibody shows ~35% cross-reactivity with recombinant mouse fractalkine/CX3CL1 and 15% cross-reactivity with recombinant rat CX3CL1. The antibody also shows 5% cross-reacitivity with recombinant human eotaxin, recombinant mouse eotaxin, recombinant human MCP-1, recombinant human MCP-2, recombinant human 6Ckine, and recombinant mouse MARC.

Immunogen

recombinant human fractalkine/CX3CL1 chemokine domain, expressed in E. coli.

Application

Anti-Fractalkine antibody can be used in western blotting and ELISA.

Physical form

Lyophilized from 0.2 μm filtered solution of phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fractalkine: one chemokine, many functions.
Gemma E White et al.
Blood, 113(4), 767-768 (2009-01-24)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service