Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F0389

Sigma-Aldrich

Fmoc-L-alanine Sasrin® resin ester

200-400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

form

solid

extent of labeling

0.3-0.8 mmol FMOC-amino acid per g resin

particle size

200-400 mesh

functional group

Fmoc

Storage temp.

2-8°C

Application

For peptide synthesis. Peptides can be cleaved from the resin with 0.5-1% CF3CO2H in CH2Cl2.

Biochem/physiol Actions

Fmoc-L-alanine is an N-terminal protected amino acid (Fmoc amino acid) used in solid-phase peptide synthesis (SPPS) to make peptides containing an the alanine residue. Fmoc-L-alanine Sasrin® (Bachem) resin ester is used for Fmoc solid phase peptide synthesis.

Other Notes

N-FMOC-amino acid ester of 3-methoxy-4-oxymethyl-phenoxymethylated 1% divinylbenzene cross-linked polystyrene.

Legal Information

Sasrin is a registered trademark of Bachem Bioscience Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service