Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F0320

Sigma-Aldrich

Fetal Bovine Serum

USA origin, Heat Inactivated, γ-irradiated, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

bovine fetus

sterility

γ-irradiated
sterile-filtered

product line

FBS Classic

form

liquid

origin

USA origin

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service