Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F0129

Sigma-Aldrich

Fibrinogen from baboon plasma

Synonym(s):

Factor I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

biological source

baboon plasma

form

solid

mol wt

α-chain 63.5 kDa
β-chain 56 kDa
γ chain 47 kDa (about 4% carbohydrate content)
soluble dimer 340 kDa

composition

Protein, ~60% (over 80% of protein clottable)

UniProt accession no.

storage temp.

−20°C

Gene Information

Biochem/physiol Actions

Acute phase protein that is part of the coagulation cascade of proteins.
Fibrinogen is an acute phase protein that is part of the coagulation cascade of proteins. The end result of the cascade is the production of thrombin that converts fibrinogen to fibrin. Thrombin rapidly proteolyses fibrinogen, releasing fibrinopeptide A. The loss of this small peptide is not sufficient to make the resulting fibrin molecule insoluble, but it tends to form complexes with adjacent fibrin and fibrinogen molecules. Thrombin then cleaves a second peptide, fibrinopeptide B, from fibrin and the fibrin monomers formed then polymerize spontaneously to form an insoluble gel. The polymerized fibrin is held together by noncovalent and electrostatic forces and stabilized by the transamidating enzyme, factor XIIIa, that is produced by the action of thrombin on factor XIII. The insoluble fibrin aggregates (clots) and aggregated platelets then block the damaged blood vessel and prevent further bleeding. The amount of fibrinogen in the plasma can serve as a nonspecific indicator of whether or not an inflammatory process is present in the body. Fibrinogen from any mammalian source will be cleaved by thrombin from any mammalian source.

Quality

Contains ~15% sodium citrate and ~25% sodium chloride.

Analysis Note

Protein determined by biuret.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P W Collins et al.
British journal of anaesthesia, 113(4), 585-595 (2014-07-30)
We aimed to create a theoretical tool to model the effect of three haemostatic agents containing fibrinogen (therapeutic plasma, cryoprecipitate, and fibrinogen concentrate) on the patient's plasma fibrinogen level. A mathematical model was developed step-wise. The relationship between the amount
Anna Agren et al.
Anesthesia and analgesia, 118(5), 933-935 (2014-05-02)
Fibrinogen is of crucial importance in patients with ongoing bleeding. In this study, we compared fibrinogen concentration measured by thrombelastography (TEG®) with fibrinogen plasma concentration determined by Clauss. Sixty-three surgical patients and 38 healthy controls were included. For the whole
Katarina Waldén et al.
The Annals of thoracic surgery, 97(4), 1199-1206 (2014-02-11)
Data from small selected patient populations suggest that the preoperative plasma concentration of fibrinogen influences postoperative blood loss and red blood cell transfusion after cardiac operations, but there are also conflicting reports. We assessed the importance of preoperative fibrinogen concentration
Jing Shi et al.
Annals of the rheumatic diseases, 73(4), 780-783 (2013-12-18)
The presence of anti-citrullinated protein antibodies (ACPA) and IgM-rheumatoid factor (IgM-RF) years before the clinical diagnosis of rheumatoid arthritis (RA) suggests they are possibly involved in the pathogenic process underlying RA. In this study, we analysed whether anti-carbamylated protein (anti-CarP)
Richard J Fish et al.
Blood, 123(14), 2278-2281 (2014-02-21)
Mutations in the human fibrinogen genes can lead to the absence of circulating fibrinogen and cause congenital afibrinogenemia. This rare bleeding disorder is associated with a variable phenotype, which may be influenced by environment and genotype. Here, we present a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service