Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ETIPLRS1000-EP

Eppendorf® epT.I.P.S.® LoRetention Reloads

PCR clean, volume × L 50-1000 μL × 71 mm, blue, pack of 960 ea (10 racks × 96 tips)

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600

material

blue

sterility

non-sterile

feature

DNase free
PCR clean
RNase free
low retention

packaging

pack of 960 ea (10 racks × 96 tips)

manufacturer/tradename

Eppendorf® 0030072030 (EU)
Eppendorf® 022493016 (NA)

technique(s)

PCR: suitable

maximum volume

1 mL

volume × L

50-1000 μL × 71 mm

foreign activity

PCR inhibitors, DNase, RNase, free

Looking for similar products? Visit Product Comparison Guide

General description

epT.I.P.S. LoRetention pipette tips and ep Dualfilter T.I.P.S. LoRetention filter tips are particularly well-suited for applications with samples that contain detergents. Detergents or detergent containing liquids lower the liquid′s surface tension. This leads to a phenomenon called “wetting”: Residual liquid sticks to the tip wall and cannot be recovered. The invisible film left behind in standard tips shows a significantly high amount of valuable sample material, which becomes discarded together with the tip.

Particularly when it comes to sensitive PCR and real-time PCR applications, low retention surfaces can increase the reproducibility and significantly reduce the loss of expensive reagents.

The low retention property of epT.I.P.S. LoRetention pipette tips and ep Dualfilter T.I.P.S. LoRetention filter tips is generated by an ultrahydrophobic, extremely homogeneous surface. It is achieved through an innovative treatment at the molecular level – the “Pearl Effect” Technology. All pipette tips are uncoated, free of additives and do not leach into the sample. Liquids containing detergents simply roll off, so that practically no liquid remains in the tip. These outstanding material properties guarantee maximum sample recovery for cost saving and improved reproducibility.

Application

  • Cell culture (media)
  • Genomics: PCR, RT-PCR, qPCR and all other types of PCR
  • Enzymatic reactions (restriction decomposition, ligation)
  • Isolation and purification of nucleic acid
  • Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton, X-100, Brij® 35, Tween® 20, CHAPS
  • Proteomics (all types of protein examinations)
  • Protein isolation and purification

Features and Benefits

  • Ultrahomogenous surface for maximum reproducibility
  • Ultrahydrophobic surface for minimal sample loss
  • Significantly reduced foam formation during pipetting
  • Extreme resistance to chemicals
  • Available in PCR clean and Eppendorf Quality
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free)
  • No coating
  • Lot-specific certificates are available at: www.eppendorf.com/certificates

Legal Information

Brij is a registered trademark of Croda International PLC
Eppendorf is a registered trademark of Eppendorf AG
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service