Key Documents
EPU9044-0006
LN2 Back-up System
for Eppendorf® CryoCube® F570 series, Premium, and HEF® freezers, AC/DC input 115 V / 208 V, 50/60 Hz, US 3-pin plug
Select a Size
1 EA
$2,330.00
$2,330.00
Check Cart for Availability
Select a Size
Change View
1 EA
$2,330.00
About This Item
UNSPSC Code:
41121800
$2,330.00
Check Cart for Availability
manufacturer/tradename
Eppendorf® U9044-0006
Related Categories
General description
Loss of power is no problem with back-up systems, which are designed to maintain the contents of your freezer at a preset temperature should a power outage arise.
Features and Benefits
Equipped with a battery back-up to temporarily protect the contents of your freezer in case of power failure
Available in either liquid CO2 and LN2. Liquid CO2 can maintain temperatures between -50 C and -70 C, where as LN2 can go down to -85 C
Both systems can be factory installed with your freezer order or retrofitted on location
Info: CO2 and LN2 are not interchangeable systems
Legal Information
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
HEF is a registered trademark of Eppendorf, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)