Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPS44I041001

Eppendorf® Innova S44i Positioning Base

base L 4.1 in. (10.16 cm)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

feature

Innova(R) S44i

packaging

pkg of 1 ea

manufacturer/tradename

Eppendorf® S44I041001

base L

4.1 in. (10.16 cm)

Features and Benefits

  • Compatible with single unit, 2x stack and 3x stack
  • For 2x stack: Stacking kit S44I041003 required in addition
  • For 3x stack: Stacking kit S44I041005 required in addition

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service