EPP0628-6260
Eppendorf® CellXpert Incubator Water Tray
for use with 170 L incubators
Synonym(s):
water reservoir
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
material
copper
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® P0628-6260
compatibility
for use with 170 L incubators
General description
The copper tray allows water to be placed at the bottom of the incubator, This acts as a heat buffer keeping the temperature more stable and prevents drastic temperature fluctuations. It also has a secondary function of ensuring proper humidity levels. The antimicrobial properties of 100% pure copper provide continuous protection against contamination on contact, all day every day.
Application
Incubator humidity
Features and Benefits
Accessory to fit 170 L Eppendorf CellExpert incubator
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service