Key Documents
EPP0628-6200
Eppendorf® Humidity Tray
0.3 L
Synonym(s):
Boxes and CA Category Containers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® (P0628-6200)
General description
Humidity tray for use in incubators to ensure samples do not dry out.
Features and Benefits
0.3 L
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)