Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPP0628-6140

Eppendorf® Humidity Tray

stainless steel, for use with 170 L incubators

Synonym(s):

water reservoir

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Eppendorf® P0628-6140

compatibility

for use with 170 L incubators

General description

The tray allows water to be placed at the bottom of the incubator, This acts as a heat buffer keeping the temperature more stable and prevents drastic temperature fluctuations. It also has a secondary function of ensuring proper humidity levels.

Application

Incubator humidity

Features and Benefits

Accessory to fit 170 L Eppendorf CellExpert incubator

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service