Key Documents
EPP0625-2110
Chart Recorder and Accessories for all Eppendorf® Freezer Models
paper, temp. range -50 — -100 °C, pack of 60
Select a Size
60 EA
$133.00
$133.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
60 EA
$133.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.12
$133.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
material
paper
packaging
pack of 60
manufacturer/tradename
Eppendorf® P0625-2110
parameter
-50 — -100 °C temp. range
Related Categories
General description
The chart recorder will continuously monitor and record freezer temperature. It is front mounted to your freezer for easy access and can be fitted to all freezer models.
Features and Benefits
  • Includes chart recorder, RTD probe, keys, chart recorder paper, and chart recorder pens
  • Will continuously monitor and record freezer temperature for data documentation
  • Front mounted to your freezer for easy access and can be fitted to all freezer models
  • Type I: Battery powered, will provide a minimum of eighteen months of operation
  • User configurable built in high and low temperature alarms
  • Can be factory installed with your freezer order at no additional charge or retrofitted on location
  • Selectable temperature range (0 to -50 C and -50 to -100 C) and rotation speed
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)