Key Documents
EPP0625-1630
TCA-3 Temperature Monitoring System
for use on use with Eppendorf® freezers, AC/DC input 120 V, 60 Hz, US 3-pin plug
About This Item
UNSPSC Code:
12352200
Product EPP0625-1630 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
Eppendorf® P0625-1630
General description
Eppendorf TCA-3 Temperature Monitoring System is an independent temperature monitoring system for those who require complete 24/7 reliable sample protection. For those that also require alarm notification, chart recorders and autodialers (country dependent) can find all these features incorporated into one small pod that integrates seamlessly to the side of your freezer. This system consists of a certified RTD thermo-probe with pod measuring 6.99 cm W x 12.55 cm D x 3.24 cm H (2.75 in W x 4.94 in D x 1.28 in H) and communicates directly through any standard Ethernet connection without the need for a computer. No additional software is required to view your collected data, all that is needed is an internet browser.
Hassel-free monitoring system for your ULT freezer
The TCA-3 system is a truly independent temperature monitoring platform for your Ultra-Low Temperature freezer. There is absolutely no software to download and you can generate reports and review alarms within minutes. This is the only system that combines a chart recorder, auto-dialer, alarm system and power monitor into one small pod. In addition, all the data collected by your TCA-3 system can be viewed from any web based computer, always keeping you informed.
Features and Benefits
  • Receive email, voicemail (country dependent), and text alarms (country dependent) if the temperature exceeds desired levels for higher sample safety
  • Quick installation on any freezer regardless of manufacturer for flexible usage
  • View temperature data readings, graphs, and statistics from any web browser for flexible data access
  • Temperature range is from -100 C to 100 C
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)