EPM1352-0004
Eppendorf® New Brunswick™ Excella® E24/E24R Incubator Shaker
E24R, refrigerated, orbit diameter 1.9 cm (3/4 in.), AC/DC input 120 V, 60 Hz
About This Item
Recommended Products
feature
CSA and CE compliant
E24R
orbit diameter 1.9 cm (3/4 in.)
refrigerated
manufacturer/tradename
Eppendorf® M1352-0004 (US)
parameter
(-15 to 60°C)
50-400 rpm (speed range)
±0.5 °C (temperature uniformity at 37°C)
W × D × H
56 cm × 76.2 cm × 61 cm
platform size
46 cm × 46 cm
weight
80 kg
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service