EPM1344-0008
Eppendorf® New Brunswick™ I24/I24R Incubator Shaker
I24, non refrigerated, orbit diameter 3/4 in., AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
About This Item
Recommended Products
feature
CSA and CE compliant
I24
non refrigerated
orbit diameter 3/4 in.
manufacturer/tradename
Eppendorf® M1344-0008 (US)
parameter
temp. range (ambient +7 to 60°C)
25-500 rpm (speed range)
W × D × H
56 cm × 76.2 cm × 61 cm
platform size
46 cm × 46 cm
weight
60 kg
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service