Key Documents
EPM1344-0004
Eppendorf® New Brunswick I24/I24R Incubator Shaker
I24R, refridgerated, orbit diameter 3/4 in., AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug, platform not included
About This Item
UNSPSC Code:
12352200
feature
CSA and CE compliant
I24R
orbit diameter 3/4 in.
platform not included
refridgerated
manufacturer/tradename
Eppendorf® M1344-0004 (US)
parameter
-15-60 °C temp. range
25-500 rpm (speed range)
W × D × H
56 cm × 76.2 cm × 61 cm
platform size
46 cm × 46 cm
weight
60 kg
General description
benchtop incubator with built-in shaker
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)