EPM1194-0010
Eppendorf® New Brunswick™ Innova® Platform Shaker
2150, size 1.9 cm , orbit diameter, AC/DC input 120 V, 60 Hz (US)
About This Item
feature
2150
CSA and CE compliant
benchtop model
platform sold separately
manufacturer/tradename
Eppendorf® M1194-0010 (US)
parameter
25-500 rpm (speed range)
W × D × H
60.1 cm × 59.1 cm × 16.3 cm
platform size
61 cm × 46 cm
size
1.9 cm , orbit diameter
weight
36 kg
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service