Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPI020

Sigma-Aldrich

Adenosine Deaminase Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

Adenosine Deaminase (ADA) (E.C. 3.5.4.4.) is an enzyme that catalyzes the conversion of adenosine and 2′-deoxyadenosine to inosine and 2′-deoxyinosine. Adenosine Deaminase is widely distributed in various tissues and cells. There are two isoforms, ADA1 and ADA2. ADA1 is widely expressed in most cells in the body, particularly in lymphocytes and macrophages. It is present in the cytosol, nucleus and found associated with dipeptidyl peptidase-4 on the cell membrane. ADA2 was first found in the spleen but is predominantly found in the plasma and serum. Increased serum ADA levels are found in certain infectious diseases such as tuberculosis and various liver diseases such as acute hepatitis, alcoholic hepatic fibrosis, chronic active hepatitis to name a few. Adenosine Deaminase is also a marker for T-lymphocyte proliferation.

Suitability

Suitable for the detection of Adenosine Deaminase activity in cell and tissue lysate, nuclear extract, cell medium and purified recombinant protein

Principle

Inosine formed from the breakdown of adenosine is detected via a multi-step reaction, resulting in the formation of an intermediate that reacts with the ADA Probe to generate a fluorescent product. The kit measures total Activity of Adenosine Deaminase with limit of quantification of 10 μU of recombinant adenosine deaminase. The range of the assay is from 10–100 μU.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service