Key Documents
EPCO48210045
Eppendorf® Galaxy® 48R CO2 Incubator
AC/DC input 120 V, parameter 1-19% (O2 control), High-Temp Disinfection
About This Item
UNSPSC Code:
41102404
AC/DC input
120 V
material
stainless steel
feature
High-Temp Disinfection
manufacturer/tradename
Eppendorf® CO48210045
parameter
1-19% (O2 control)
4-50 °C temp. range
General description
The Galaxy 48 R CO2 Incubator is a mid-sized, 48 liter incubator designed to optimize performance in applications that require a higher level of isolation and can be used in small labs. The 48 R series of incubators feature a large backlit display with on screen help menu, control and data logging of temperature, CO2, door openings, alarms, and optional O2 gas and humidity monitoring.
Features and Benefits
  • 48 Liter (1.7 ft3) provides mid-sized capacity within a small footprint
  • Options: High Temp. Disinfection (4 hour at 120 °C), O2 control (e.g. for hypoxic studies), 2-split inner glass door
  • Fanless design offers more usable space
  • Six-sided direct heating system provides a uniform incubated environment
  • Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
  • Low gas consumption
  • Integrated alarm relay
benchtop, under bench, floor, or stackable (x2)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Galaxy is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)