Key Documents
EP950014016
CAN Bus Connecting Cable, for Eppendorf® Mastercycler® Nexus and Pro Thermal Cyclers
L 150 cm
About This Item
UNSPSC Code:
41121800
manufacturer/tradename
Eppendorf® 950014016 (North America)
L
150 cm
General description
  • Designed for use with Eppendorf Mastercycer pro thermal cyclers
  • Allows for data connectivity between multiple thermal cyclers and PC control software
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)