Key Documents
EP950014008
CAN Bus Connecting Cable, for Eppendorf® Mastercycler® Nexus and Pro Thermal Cyclers
L 50 cm
Select a Size
1 EA
$58.00
$58.00
Available to ship onApril 29, 2025Details
Select a Size
Change View
1 EA
$58.00
About This Item
UNSPSC Code:
41121800
$58.00
Available to ship onApril 29, 2025Details
manufacturer/tradename
Eppendorf® 950014008 (North America)
L
50 cm
Related Categories
General description
  • Designed for use with Eppendorf Mastercycer pro thermal cyclers
  • Allows for data connectivity between multiple thermal cyclers and PC control software
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)