Key Documents
EP6135000923
Eppendorf BioSpectrometer® Basic
basic, INCLUDING μCUVETTE® G1.0, AC/DC input 120 V
About This Item
feature
INCLUDING μCUVETTE® G1.0
basic
manufacturer/tradename
Eppendorf® 6135000923
General description
The Eppendorf BioSpectrometer basic is a small, very compact spectrophotometer that combines easy handling with highly reproducible analysis. It can measure and record UV/Vis spectral ranges as well as measure individual wavelengths from 200 nm to 830 nm. Its freely selectable wavelengths provide maximum flexibility for all current and future applications. In addition to preprogrammed methods for standard measurements, users can create and store their own methods. Optimized menu navigation guides through the individual methods in a step-by-step process. All required entries are visible, no essential data is omitted. A help box explains the operational sequences and is available in 5 different languages.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf BioSpectrometer is a registered trademark of Eppendorf AG
Eppendorf µCuvette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)