EP5942000741
Eppendorf® Centrifuge 5910R G, refrigerated
incl. S-4x400 rotor, round buckets and adapter for conical tubes, 5/15/50mL, AC/DC input 120 V AC, US 3-pin plug
About This Item
Recommended Products
feature
CSA approved
refrigerated
to hold 16 50 mL conical tubes
to hold 28 15 mL conical tubes
to hold 4 400 mL round buckets
incl. S-4x400 rotor, round buckets and adapter for conical tubes, 5/15/50mL
manufacturer/tradename
Eppendorf® 5942000741
parameter
temp. range at -11-40 °C
13700 rpm
21194 × g
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 750 mL
- Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
- Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
- High centrifugation speed of up to 22,132 x g (14,000 rpm)
- Aerosol-tight Eppendorf QuickLock caps and lids allow fast and ergonomic lid locking
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service