EP5942000315
Eppendorf® Centrifuge 5910 R G
refrigerated, incl. S-4xuniversal rotor, universal buckets and adapter for 5/15/50 mL conical tubes and plates, AC/DC input 230 V, 50 - 60 Hz
About This Item
Recommended Products
AC/DC input
230 V AC, Schuko plug
feature
CE compliant
refrigerated
to hold 36 50 mL conical tubes
to hold 68 15 mL conical tubes
incl. S-4xuniversal rotor, universal buckets and adapter for 5/15/50 mL conical tubes and plates
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 5942000315 (EU)
parameter
temp. range at -11-40 °C
14000 rpm
22132 × g
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 750 mL
- Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
- Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
- High centrifugation speed of up to 22,132 × g (14,000 rpm)
- Aerosol-tight Eppendorf QuickLock caps and lids allow fast and ergonomic lid locking
- Centrifuge lid with soft-touch lid closure
- Quiet operation to improve your work environment
- Compact footprint saves valuable bench space
- Automatic rotor recognition and imbalance detection for maximum operational safety
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service