Key Documents
EP5804730003
Eppendorf® Drum Rotor
T 60-1, incl. lid, for use with 6 adapters
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5804730003 is not currently sold in your country. Contact Technical Service
feature
to hold 60 x 1.5/2.0 mL tubes
T 60-1, incl. lid, for use with 6 adapters
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5804730003
parameter
14,000 max. RCF (16,435 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)