Key Documents
EP5492008003
Eppendorf® Swing-bucket-rotor A-8-11
8,1.5/2 ml, incl. aluminum lid
About This Item
UNSPSC Code:
41103909
Product EP5492008003 is not currently sold in your country. Contact Technical Service
feature
to hold 8 x 1.5/2.0 mL vessels
8,1.5/2 ml, incl. aluminum lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5492008003
parameter
10,000 max. RCF (10,600 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5417C/R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)