Key Documents
EP5490061004
Eppendorf® Fixed-angle rotor
FA-45-24-11, 24 x 1,5/2 ml, incl. aluminium lid, aerosol-tight, f. 5417C/R
About This Item
UNSPSC Code:
41103909
Product EP5490061004 is not currently sold in your country. Contact Technical Service
feature
to hold 24 x 2.0 mL tubes
FA-45-24-11, 24 x 1,5/2 ml, incl. aluminium lid, aerosol-tight, f. 5417C/R
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490061004
parameter
16,400 max. RCF (25,000 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5417C/R
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)