Key Documents
EP5427705007
Eppendorf® Rotor F-45-18-17-Cryo
incl. rotor lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5427705007 is not currently sold in your country. Contact Technical Service
feature
to hold 18 x cryovials
incl. rotor lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427705007
parameter
11,800 max. RCF (8,324 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)