Key Documents
EP5425737009
Eppendorf® Fixed-angle rotor
FA-45-24-11, aluminium, cpl. with aluminium lid, aerosol-tight
About This Item
UNSPSC Code:
41103909
feature
to hold 24 x 2.0 mL tubes
FA-45-24-11, aluminium, cpl. with aluminium lid, aerosol-tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5425737009
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5415D/R
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)