Key Documents
EP5425725000
Eppendorf® Standard Rotor F-45-24-11
About This Item
UNSPSC Code:
41103909
feature
to hold 24 x 2.0 mL tubes
aluminium, cpl. w. polypropylene lid, non aerosol-tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5425725000
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5415D/R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)