Key Documents
EP5404000060
Eppendorf® Centrifuge 5424 R G
refrigerated, AC/DC input 230 V, 50 - 60 Hz, foil keypad version, incl. rotor FA-45-24-11, aerosol-tight, AC/DC input, UK plug
About This Item
UNSPSC Code:
40161701
AC/DC input
230 V AC, UK plug
feature
CE compliant
to hold 24 1.5/2.0 mL tubes
foil keypad version, incl. rotor FA-45-24-11, aerosol-tight
refrigerated
manufacturer/tradename
5404000060
Eppendorf® 5404000060
Eppendorf®
parameter
temp. range at -10-40 °C
15000 rpm
21130 × g
technique(s)
centrifugation: suitable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)