Key Documents
EP5195000087
Eppendorf® Piezo Drill Tip
(M. ICSI), sterile
About This Item
UNSPSC Code:
41121600
sterility
sterile
feature
(M. ICSI)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 5195000087
General description
  • Piezo Drill Tip designed for cell manipulation application.
  • 6 μm inner diameter.
  • 25° tip angle for optimal pulse transfer.
  • 6 mm flange length.
  • Non-polished, blunt tip end for optimal piezo pulse force transfer and minimal trauma.
  • Sterilized via validated gamma irradiation.
  • Capillary safe for optimum protection.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)