Key Documents
EP5195000044
Eppendorf® VacuTip II
sterile, set of 25 ea
About This Item
UNSPSC Code:
41121600
sterility
sterile
packaging
set of 25 ea
manufacturer/tradename
5195000044
Eppendorf®
General description
  • 60 μm inner diameter
  • 110 μm outer diameter
  • 25° tip angle
  • Flange length 0.5 mm
  • Smooth rounded tip end for gentle holding
  • Sterilized via validated gamma irradiation
  • Non-cytotoxic (as proven by mouse embryo assay)
  • Capillary safe for optimum protection
Application
Eppendorf® VacuTip II is useful for holding of oocytes or blastocytes.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)