Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5195000036

Eppendorf® VacuTip I

sterile, set of 25 ea

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600

sterility

sterile

packaging

set of 25 ea

manufacturer/tradename

5195000036
Eppendorf®

General description

  • 15 μm inner diameter
  • 100 μm outer diameter
  • 35° tip angle
  • Flange length 1 mm
  • Smooth, rounded tip end for gentle holding
  • Sterilized via validated gamma irradiation
  • Non-cytotoxic (as proven by mouse embryo assay)
  • Capillary safe for optimum protection

Application

Eppendorf® VacuTip II is useful for holding of oocytes or blastocytes.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service