Key Documents
EP4966511007
Ventilation plugs for Eppendorf® Varispenser® 2 / 2x
with Luer cone made of PP, including PTFE sealing ring, all sizes
Select a Size
1 EA
$84.00
$84.00
Check Cart for Availability
Select a Size
Change View
1 EA
$84.00
About This Item
UNSPSC Code:
41121517
NACRES:
NB.01
$84.00
Check Cart for Availability
feature
including PTFE sealing ring, all sizes
with Luer cone made of PP
manufacturer/tradename
Eppendorf® 4966511007
General description
Ventilation screw for microfilters with Luer connection. Made from polyproplyene with a PTFE washer.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Varispenser is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)