EP2006
Sigma-Aldrich® power output adapter
2 mm to 4 mm adapter
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
packaging
pack of 2 (cables)
Application
Useful for connecting different brands of electrophorisis units and power supplies
Recommended products
Also see power supplies and power cables for related products
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)