Key Documents
EP2001
Sigma-Aldrich
Sigma-Aldrich® power supply
CS-300V, AC/DC input 110 - 230 V AC
About This Item
UNSPSC Code:
41100000
description
EU, UK and US power cords included
feature
CE compliant
  • Ability to store up to 10 methods
  • Timer range: 1-999 minutes
manufacturer/tradename
CS-300V
output
300 V, 700 mA, 150 W
Biochem/physiol Actions
Can be set up to run in constant voltage and constant current modes. Able to connect 4 gel chambers. Optional timer can be set to end a run and sound an alarm.
Caution
Safety features include shrouded plugs and sockets, no load detection, and ground leakage detection.
Recommended products
Also see power cables and power output adaptors for accessory products.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)