Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP1501

Sigma-Aldrich

Sigma-Aldrich® SB10 omniPAGE mini electroblotting system

10x 10cm, with 4 cassettes and 8 fibre pads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NC.01

General description

A stand-alone complete unit for electroblotting. Includes tank and lid, electroblotting insert, cassettes, and fibre pads. Hinged rigid cassettes allow rapid set up and convenience and ensures even contact between the gel and membranes.

Application

Ideal for western blotting applications..Open design and rigid cassettes offer maximum current transfer.

Components

System includes 4 cassettes and 8 fibre pads

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service