Key Documents
EP1401
Sigma-Aldrich
Sigma-Aldrich® VS20WAVESYS omniPAGE maxi dual vertical electrophoresis system
20x20 cm, with caster
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
grade
electrophoresis grade
W × D × H
26 cm × 16 × 28 cm
volume × volume
min 1200 mL × max 5600 mL
General description
The preferred unit for maxi protein electrophoresis, the omniPAGE maxi unit combines convenient ease of use features with high resolution separations. Simple set up using ultra soft silicone seals guarantees trouble free glass plate loading and gel casting. Dual gaskets on the gel running insert along with notched and plain glass plates ensure leak proof gel running. Rapid setup cooling retains resolution in extended separations and also saves on buffer volume without affecting run quality. 4 mm thick glass plates reduce breakage and have bonded spacers for added convenience. A wide range of accessories are available allowing many techniques to be performed using the same unit. Prep combs can be used to maximize sample loading and recovery. Accessory electroblotting and tube gel modules are available which use the same outer tank and lid.
  • Ideal for high resolution separations
  • Typical applications include Maxi SDS PAGE, Native PAGE, Gradient, Second dimension and Nucleic acid separations
  • Rapid gel casting and loading
  • Optional low or high buffer volumes
  • Rapid set up cooling
Included with the omniPAGE maxi unit are dual 20 x 20 cm glass plates with bonded 1 mm thick spacers, 2 x 24 sample combs, cooling pack, dummy plate and Casting Base. Furthermore, these units can accommodate gels with the following dimensions (W x L): 16 x 17.5 cm. In addition, this unit accommodates a variety of comb options that include sample options of 1, 5, 10, 18MC, 24, 30, 36MC and 48 as well as comb featuring 0.75, 1, 1.5 and 2.0 mm thickness.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)