Key Documents
EP1351
Sigma-Aldrich
Sigma-Aldrich® CVS10 comb
10 sample, thickness 1.0 mm
About This Item
UNSPSC Code:
41100000
thickness
1.0 mm
well volume
40 μL
General description
Comb Prep number indicates the number of preparatory wells. Marker number indicates the number of small volume wells intended for molecular weight markers. MC indicates multichannel pipette compatible.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)