Key Documents
EP1322
Sigma-Aldrich
Sigma-Aldrich® CVS10 glass plate
size 10 cm × 10 cm , notched, with bonded spacers, thickness 1.5 mm, pack of 2 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
packaging
pack of 2 ea
size
10 cm × 10 cm , notched, with bonded spacers
thickness
1.5 mm
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)